Filtration crucibles, jacketed, VitraPOR®
Leverancier: ROBU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VitraPOR®
ROBU49903EA 748 EUR
ROBU49903 ROBU49902 ROBU49704 ROBU49703 ROBU49901 ROBU49702 ROBU49900 ROBU49701 ROBU49304 ROBU49700 ROBU49303 ROBU49302 ROBU49904 ROBU49301 ROBU49300
Filtration crucibles, jacketed, VitraPOR®
Kroezen
The crucible is made of pure borosilicate glass 3.3, versatile and recyclable material offers excellent properties for the most diverse fields.
  • Porosity classes ranges from 1,0 µm to 500 µm
  • Contains heating mantle and standard joints
  • Highly resistant to most chemicals such as solvents, acids and alkalis