Chromocult® listeria-selectieve agar
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.00427.0500EA 407 EUR
1.00427.0500
Chromocult® listeria-selectieve agar
Media voor microbiologie Gedehydrateerde media voor microbiologie
This medium conforms to the ALOA (agar Listeria acc. to Ottaviani and Agosti) medium, a selective medium for the isolation of Listeria monocytogenes in food (AFNOR-validated method). It also enables the testing, sampling and enumeration of Listeria monocytogenes and Listeria spp. in foodstuffs and other source materials. The rich base of the Chromocult® Listeria selective agar encourages excellent growth. The supplement of inhibitors stops most of the bacteria, yeast and fungi from growing. In contrast, Listeria is uninhibited in its growth, producing blue-green colonies; L. monocytogenes also form an opaque halo.
  • Eenvoudige, veilige interpretatie
  • ISO 11290 Referentievoedingsmedium, door AFNOR gevalideerd
  • Door FDA/BAM aanbevolen