Netherlands
Order Entry
M-FIX™ Spray fixative for gynecological and clinical cytology, Sigma-Aldrich®
M-FIX™ Spray fixative for gynecological and clinical cytology, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
M-FIX™ Spray fixative for gynecological and clinical cytology, Sigma-Aldrich®
Supplier:  Merck
Related Information
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Spray fixative for cytodiagnosis
    Spray fixative for cytodiagnosis
    1.03981.0102
    1.03981.0102
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 20,50 €
    20,50 €
    1 * 100 mL
       20,50 €
    Supplier Article Number: 1.03981.0102
    Local Article Number: MERC1.03981.0102
  • Spray fixative for cytodiagnosis
    Spray fixative for cytodiagnosis
    1.03981.1000
    1.03981.1000
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 181,00 €
    181,00 €
    1 * 1 L
       181,00 €
    Supplier Article Number: 1.03981.1000
    Local Article Number: MERC1.03981.1000