Desiccator cabinets, auto, vertical, secador®, SP Bel-Art
Leverancier: Bel-Art Products, a part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Secador®
BELAF420731220EA 2430 EUR
BELAF420731220 BELAF420741228 BELAF420721228 BELAF420741226 BELAF420741227 BELAF420741220 BELAF420721220
Desiccator cabinets, auto, vertical, secador®, SP Bel-Art
Exsiccatoren Exsiccatorkasten
Secador® model 2.0, 3.0 and 4.0 auto desiccator cabinets are made from moulded Durastar® co-polyester which blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Large door with easy-to-view digital hygrometer
  • Perforated shelves facilitate circulation of dry air
  • Colour-indicating silica gel allows quick visual confirmation of desiccant status
  • Amber colour units reduce visible light penetration by over 50%; perfect for storing light sensitive materials like reagents, analytical standards and investigational compounds
Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® Auto-Desiccator cabinets. Automatic dehumidification and desiccant regeneration cycle operates continuously every 20 minutes. Compared to other auto-desiccators that regenerate once every 4 to 6 hours, the Secador® Auto-Desiccators attain a low %rH level on start-up and consistently maintain low %rH, especially with frequent entry to the desiccator interior.
Leveringsinformatie: Supplied with power cord/transformer with input connector.