Face shields
Leverancier: Uvex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
111-1577EA 63.3 EUR
111-1577 UVEX9708.514
Face shields
Gelaatsschermen
Robust visor with integrated forehead cover, clear cellulose acetate lens and additional protective cap made from impact-resistant PS.
  • Twist ratchet enables simple and reliable adjustment to any head size
  • Flip-up mechanism allows the visor to be set in any position
  • Easy lens replacement
Certificaten: EN 166