Scraper, Chisel-type, SP Bel-Art
Leverancier: Bel-Art
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BELAH368190000EA 94.6 EUR
BELAH368190000
Scraper, Chisel-type, SP Bel-Art
Schrapers
Rigid plastic scraper with flat PS blade (59 mm wide).
  • Tapered, sharp edge is slightly curved
  • Made of FDA grade materials for direct contact with foods, drugs, and cosmetics
  • Side edges are rounded