Tubing couplings, POM, NW 6,4 mm
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BURK8782-1040EA 2.9 EUR
BURK8782-1040 BURK8782-1060 BURK8783-1062 BURK8783-1060 BURK8781-1040 BURK8780-1060 BURK8780-1040 BURK8781-1060
Tubing couplings, POM, NW 6,4 mm
Fittingen en connectoren
POM, white/gray.
  • Very good chemical resistance
  • Very good temperature resistance
  • Secure O-ring guarantees that rapid couplings are leakproof
Fast and safe connection for liquids, gases, compressed air and vacuum. Rapid couplings are the ideal when connecting and separating hoses, machines, apparatus, containers, and more.