Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
U heeft gezocht op: "antibiotica-en-antimycotica"
https://nl.vwr.com/stibo/smallweb/std.lang.all/49/00/18944900.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"157"
  List view Grid view Easy View (new)
Sorteer op:
Catalogus nummer: (1.05177.0005)
Leverancier:  Merck
Omschrijving:   KANAMYCINEDISULFAAT BIOCHEM 1 * 5 g
UOM:  1 * 5 g
Leverancier:  VWR Chemicals
Omschrijving:   Kanamycin is an aminoglycoside antibiotic that inhibits protein synthesis in gram-negative and gram-positive bacteria and in mycoplasma. Recommended w...
UOM:  1 * 20 ml
Catalogus nummer: (J607-100MG)
Leverancier:  VWR Chemicals
Omschrijving:   An aminoglycoside antibiotic that inhibits protein synthesis in bacteria, fungi and higher eucaryotic cells. Widely used as a selection agent for proc...
UOM:  1 * 100 mg
Catalogus nummer: (J847-20ML)
Leverancier:  VWR Chemicals
Omschrijving:   Used as a selection agent for both prokaryotic- and eukaryotic-transfected cells. An aminoglycoside similar to gentamycin, G418 is toxic to bacterial,...
UOM:  1 * 20 ml
Leverancier:  VWR Chemicals
Omschrijving:   Aseptic, gamma-irradiated, tissue culture tested. 10 000 U/ml penicillin. 10 mg/ml streptomycin solution when reconstituted in 20 ml of sterile w...
UOM:  1 * 20 ml
Catalogus nummer: (K547-20ML)
Leverancier:  VWR Chemicals
Omschrijving:   An aminoglycoside antibiotic that inhibits protein synthesis in bacteria, fungi and higher eucaryotic cells. Inhibits chain elongation. Widely used as...
UOM:  1 * 20 ml
Catalogus nummer: (E737-20ML)
Leverancier:  VWR Chemicals
Omschrijving:   Gefilterd door een 0,22 µm filter. 15 µg/ml werkende concentratie, bindt zich aan de 30S subunit van bacteriële ribosomen.
UOM:  1 * 20 ml
Leverancier:  VWR Chemicals
Omschrijving:   Pennicillin-Streptomycin 100X sterile solution for tissue culture applications. Inhibits cell wall synthesis and protein sythesis in gram-positive and...
UOM:  1 * 100 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 157
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Volgende