Denhardt's solution, 100X, for biotechnology
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E257-50MLEA 91.3 EUR
VWRCE257-50ML
Denhardt's solution, 100X, for biotechnology
A blocking reagent for Northern and Southern blots that prevents probe binding on open membrane space and non target nucleic acids.