DNA band excisor, SP Bel-Art
Leverancier: Bel-Art Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BELAF369700010EA 62.5 EUR
BELAF369700010
DNA band excisor, SP Bel-Art
Celschrapers
For safely excision of DNA bands, this double ended polystyrene tool with thin blade and spatula is perfect for excising bands from agarose or polyacrylamide gels with precision and placing into test tubes.
  • Excise DNA bands as well as purified plasmid DNA preps from electrophoresis gels
  • Polystyrene tool will not cut fingers or scratch transilluminator screens
  • Speeds up processing time, reducing exposure of nucleic acids and user's hands to damaging UV light
  • Discard in standard waste receptacles; no sharps container required