Dial caliper, SP Bel-Art
Leverancier: Bel-Art Products, a part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BELAH134160001EA 327 EUR
BELAH134160001
Dial caliper, SP Bel-Art
Schuifpassers
This dial caliper can measure up to 150 mm on a white dial graduated in 0,1 mm increments. Vernier model measures 130 mm in 0,05 subdivisions, plus 5" scale with 0,001 subdivisions.
  • Adjustable dial for zero settings and differential measurements
  • Easy-to-use thumb roller adjusts jaws to measure internal and external diameters, and depth
Designed to measure glassware and plasticware accurately and safely, with durable plastic polymer.