Anemometer, testo 416
Leverancier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TEST0560.4160EA 492 EUR
TEST0560.4160
Anemometer, testo 416
windmeters
The Testo 416 is an easy to use small vane anemometer with an attached vane probe on a telescoping handle to measure CFM in air ducts. This handheld digital vane anemometer allows you to switch between volume flow and velocity readings quickly.
  • Timed average eases and quickens the task of duct register traverses
  • Hold button to freeze readings
  • Optional Topsafe case provides superior protection from dirt, moisture, and impact
  • Auto-off function saves battery life
Timed and multi-point mean flow calculations allow easy flow analysis, volume accumulation estimates, and temperature variation monitoring. Min-Max values are one button simple, while the 'Hold' function freezes the current readings in the display for easy data recording.
Leveringsinformatie: Supplied with 416 small vane anemometer, telescoping handle probe, batteries and calibration certificate.