Anemometer, testo 425
Leverancier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TEST0560.4251EA 490 EUR
TEST0560.4251
Anemometer, testo 425
windmeters
The Testo 425 hot wire anemometer provides incredible accuracy and is easy to use. The bright backlit display provides easy viewing in any environment. The telescopic handle of this meter extends to 838 mm, making even high-up ductwork within reach.
  • Compact, thermal sensor is ideal for in-duct readings
  • Measure air velocity from 0 to 4000 FPM
  • Telescoping hot wire probe
  • Velocity, volume flow (CFM) and temperature
  • Timed and multi-point average flow calculations
  • Auto-off function saves battery life
Leveringsinformatie: Supplied with 425 hot wire anemometer, batteries and calibration protocol.