Earmuffs, Peltor™ Optime™ PTL
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MMMAPTLAEA 54.6 EUR
MMMAPTLA
Earmuffs, Peltor™ Optime™ PTL
Peltor™ Optime™ Push To Listen (PTL) earmuffs are fitted with a simple sound reproduction facility to enable better speech intelligibility when used in a noisy background. By simply pressing a button located on the outside of the cup, the product allows you to listen to important speech conversation against a noisy background. This feature helps facilitate the need for communication without having to remove the earmuffs from the ears.
  • Hearing protection with manual listening function
  • Electronic function can last up to 30 seconds from time of activation, but can be de-activated at any time during the 30 second period
  • Liquid filled sealing rings,‎ 2-point attachment
Ideal for use on construction sites and on airports.
SNR: 31 dB
Certificaten: EN 352-1