Overalls with Bib and Braces, Fristads® PR54-22, Design B, Royal Blue
Leverancier: Kwintet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Fristads®
113-02233EA 0 EUR
113-02233 113-02234 113-02198 113-02231 113-02232 113-02199 113-02196 113-02197 113-02230 113-02194 113-02195 113-02239 113-02237 113-02238 113-02235 113-02236 113-02192 113-02193 113-02190 113-02191 113-02244 113-02245 113-02242 113-02243 113-02240 113-02241 113-02248 113-02249 113-02246 113-02247 113-4765 113-4766 113-4767 113-4800 113-4801 113-4768 113-4763 113-4764 113-02255 113-02256 113-02253 113-02254 113-02251 113-02252 113-02250 113-02259 113-02257 113-02258 113-02145 113-02146 113-02264 113-02262 113-02263 113-02260 113-02261 113-02149 113-02147 113-02148 113-02156 113-4828 113-4829 113-02157 113-02154 113-02155 113-4824 113-02152 113-02153 113-4825 113-4826 113-02150 113-4827 113-02151 113-02158 113-02159 113-4790 113-4791 113-4792 113-4793 113-4798 113-4831 113-4799 113-4832 113-4833 113-4834 113-4794 113-4795 113-4796 113-4797 113-4830 113-02167 113-02200 113-02168 113-02201 113-02165 113-02166 113-02163 113-02164 113-02161 113-02162 113-02208 113-02209 113-02206 113-02207 113-02204 113-02205 113-02169 113-02202 113-02203 113-02160 113-4806 113-02178 113-02211 113-02179 113-02212 113-4807 113-4808 113-02176 113-4809 113-02210 113-02177 113-02174 113-4769 113-4802 113-4803 113-02175 113-02172 113-4804 113-4805 113-02173 113-02219 113-02217 113-02218 113-02215 113-02216 113-02213 113-02214 113-4770 113-4771 113-02170 113-4776 113-02171 113-4777 113-4810 113-4778 113-4811 113-4812 113-4779 113-4772 113-4773 113-4774 113-4775 113-02189 113-02222 113-4817 113-4818 113-02223 113-4819 113-02220 113-02187 113-02188 113-02221 113-4813 113-02185 113-4814 113-02186 113-4815 113-02183 113-4816 113-02184 113-02228 113-02229 113-02226 113-02227 113-02224 113-02225 113-4780 113-4781 113-4782 113-4820 113-4787 113-02181 113-02182 113-4788 113-4821 113-4789 113-4822 113-02180 113-4823 113-4783 113-4784 113-4785 113-4786
Overalls with Bib and Braces, Fristads® PR54-22, Design B, Royal Blue
Werkkleding
Overalls made from 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two chest pockets, one with flap
  • Two front pockets, two back pockets
  • Leg pocket with flap, mobile phone pocket, pen pocket, folding-ruler pocket
  • Adjustable waist, side openings
Choice of many different accent colours.
Weight: 260 g/m²
Basic colour: Royal Blue