Overalls with Bib and Braces, Fristads® PR54-22 Design B, black
Leverancier: Kwintet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Fristads®
113-02475EA 0 EUR
113-02475 113-02476 113-02473 113-02474 113-02471 113-02472 113-02470 113-02479 113-02477 113-02478 113-02486 113-02487 113-02484 113-02485 113-02482 113-02483 113-02480 113-02481 113-02406 113-02407 113-02404 113-02405 113-02402 113-02403 113-02488 113-02400 113-02401 113-02489 113-02497 113-02410 113-02498 113-02495 113-02496 113-02493 113-02494 113-02491 113-02492 113-02417 113-02418 113-02415 113-02416 113-02413 113-02414 113-02499 113-02411 113-02412 113-02408 113-02409 113-02490 113-02387 113-02420 113-02388 113-02421 113-02385 113-02386 113-02428 113-02429 113-02426 113-02427 113-02424 113-02425 113-02422 113-02389 113-02423 113-02419 113-02431 113-02398 113-02432 113-02399 113-02396 113-02397 113-02430 113-02394 113-02395 113-02392 113-02393 113-02439 113-02437 113-02438 113-02435 113-02436 113-02433 113-02434 113-02390 113-02391 113-02442 113-02443 113-02448 113-02449 113-02446 113-02447 113-02444 113-02445 113-02453 113-02454 113-02451 113-02452 113-02450 113-02459 113-02457 113-02458 113-02455 113-02456 113-02464 113-02465 113-02462 113-02463 113-02460 113-02461 113-02503 113-02504 113-02501 113-02468 113-02469 113-02502 113-02466 113-02500 113-02467
Overalls with Bib and Braces, Fristads® PR54-22 Design B, black
Werkkleding
Thanks to their durability and comfort, these trousers are perfectly suited to workshops, and industrial and service enterprises. The material is made from 65% polyester and 35% cotton. The inside is brushed, ensuring a pleasantly comfortable cotton feel. The outside can withstand the roughest demands and will not fade, even after countless washes.
  • Two breast pockets, one with flap
  • Two front and back pockets, leg pocket with flap, mobile phone pocket, pen pouch, folding-rule pocket
  • Adjustable waist, slit opening, plastic press studs
  • Colour contrast: Slit edges, stripes on the bib and leg pocket flap
Weight: 260 g/m²
Basic colour: Black