Wool socks, Match
Leverancier: Kwintet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Match
KWIN100622940SEA 0 EUR
KWIN100622940S KWIN100622940L KWIN100622940M
Wool socks, Match
Werkkleding
Durable socks made of 80% wool, 15% polyamid and 5% Lycra.
  • Specially developed woollen fabric
  • Keeps feet dry and warm