In-line refractometer, PRM-100α
Leverancier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ATAG3574EA 0 EUR
ATAG3574
In-line refractometer, PRM-100α
In-line refractometer designed for the continuous measurements of refractive index, user-defined concentration and Brix scale. Light refraction is calculated between the prism on the detection section and the liquid running through the pipe.
  • Wider Brix range
  • Easy to read colour display panel
  • RS-232C output for connection to a computer
  • Can be mounted on the pipe or on a by-pass
Process refractometers are suitable for use in food, beverage and pharmaceutical plants as well as for control concentrations of industrial solutions (cutting oils, quenches, washes).
Data outputs allow to automatically control mixing ratios.
Bestelinformatie: A calibration certificate is available for each unit - Please contact your Avantor local agency for more information.