U heeft gezocht op: Antibioticatesten
12  results were found
SearchResultCount:"12"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (1.11499.0001)
Leverancier: Merck
Omschrijving: Voor de test op antibiotische sulfonamide-residuen volgens Kundrat. De Geobacillus stearothermophilus sporensuspensie wordt gebruikt in combina...
UOM: 1 * 5 Ampul
MSDS
Catalogus nummer: (1.10649.0001)
Leverancier: Merck
Omschrijving: Bacillus subtilis (BGA) sporensuspensie wordt gebruikt in combinatie met de testagar voor voedingsbodems pH 6,0 voor de inhibitortest art.nr. 1...
UOM: 1 * 15 Ampul
MSDS
Leverancier: THERMO FISHER DIAGNOSTICS GMBH
Omschrijving: Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliab...
Leverancier: LIOFILCHEM S.R.L.
Omschrijving: Antibiotic Tests, Ceftaroline Cpt 30 µg
Leverancier: THERMO FISHER DIAGNOSTICS GMBH
Omschrijving: Used as a screening test prior to serological grouping.
Leverancier: THERMO FISHER DIAGNOSTICS GMBH
Omschrijving: 25 discs/vial.
Leverancier: Whatman products (Cytiva)
Omschrijving: Antibiotic Tests, Whatman™, Antibiotica-assay-schijven, Ø 13 mm
Leverancier: THERMO FISHER DIAGNOSTICS GMBH
Omschrijving: Place antimicrobial susceptibility disks onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-...
Leverancier: LIOFILCHEM S.R.L.
Omschrijving: Multodisc with eight antimicrobial agents on a ring.
Leverancier: MACHEREY-NAGEL
Omschrijving: These products are used in testing the resistance of pathogens to antibiotics. For this test the filter paper sections can be impregnated with the ant...
Leverancier: PRAT DUMAS
Omschrijving: For determining antibiotic suited to the treatment of an infection and for checking their sensitivity to antibiotics by means of the inhibition zone d...
Leverancier: DIFCO (PART OF BD)
Omschrijving: Antibiotic Tests, Cefinase Paper
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 12 of 12
no targeter for Bottom