Flasks, pear shaped, with two necks
Leverancier: Witeg
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WITG0.663.050EA 32.45 EUR
WITG0.663.050 WITG0.663.025 WITG0.663.250.E WITG0.663.100
Flasks, pear shaped, with two necks
Kolven Peervormige kolven
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.