U heeft gezocht op: Microarray instrumenten
4  results were found
SearchResultCount:"4"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Molecular Devices
Omschrijving: The GenePix® 4300A and GenePix® 4400A scanners offer high resolution, high quality imaging with 3-laser excitation. Configurations include 5 μm or 2,5...
Catalogus nummer: (MLDVGENEPIX4100A-U)
Leverancier: Molecular Devices
Omschrijving: GenePix® 4100A Microarray Scanner offers its simplicity, reliability and flexibility in microarray-based research, be it in genomics, proteomics, or n...
UOM: 1 * 1 ST
Catalogus nummer: (MLDVGENEPIX4000B-U)
Leverancier: Molecular Devices
Omschrijving: The GenePix® 4000B microarray scanner is a benchmark for quality, reliability and ease-of-use in microarray scanning technology. Coupled with GenePix®...
UOM: 1 * 1 ST
Leverancier: Simport Scientific
Omschrijving: T-Sueâ„¢ Microarray mold, enabling tissues from many donor blocks to be arrayed on a single slide.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 4 of 4
no targeter for Bottom