DNA extraction kits, ZyGEM™ prepGEM® bacteria kit
Leverancier: MicroGEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ZyGEM™ prepGEM®
ZYGEPBA0500EA 1880 EUR
ZYGEPBA0500
DNA extraction kits, ZyGEM™ prepGEM® bacteria kit
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents
prepGEM® DNA extraction kits have been developed to provide a rapid, reliable, and economical solution to DNA extraction. The kits use a single tube protocol with temperature control to prepare DNA for analysis, and are optimised for downstream PCR-based applications, including qPCR, STR-PCR, DNA sequencing, AFLP analysis, and many SNiP detection applications.
prepGEM® Bacteria kit is a single tube, enzymatic DNA extraction system specifically optimised for the extraction of DNA from a wide variety of gram-negative and gram-positive bacteria. The kit uses sequential enzymatic treatment to lyse the cells and prepare the DNA template in conjunction with a specific wash and extraction buffer. The kit has been validated on more than 30 species of bacteria and typically delivers DNA yields sufficient for multiple PCRs in less than 25 minutes. The extracted DNA is suitable for PCR, qPCR, STR-PCR, or WGA and has been tested on colonies grown on agar, liquid bacterial cultures, and biomedical swab sample formats.
prepGEM® Bacteria kit is a single tube, enzymatic DNA extraction system specifically optimised for the extraction of DNA from a wide variety of gram-negative and gram-positive bacteria. The kit uses sequential enzymatic treatment to lyse the cells and prepare the DNA template in conjunction with a specific wash and extraction buffer. The kit has been validated on more than 30 species of bacteria and typically delivers DNA yields sufficient for multiple PCRs in less than 25 minutes. The extracted DNA is suitable for PCR, qPCR, STR-PCR, or WGA and has been tested on colonies grown on agar, liquid bacterial cultures, and biomedical swab sample formats.