Conductivity electrode, four-pole
Leverancier: Consort
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CONSSK40TEA 410 EUR
CONSSK40T
Conductivity electrode, four-pole
Elektroden Geleidbaarheidselektroden
This four graphite electrode with integrated temperature sensor permits to measure low and high conductivites with only one electrode. For use with meter K912.