Spreaders, L-shaped, Sterilin®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
391-0480EA 348 EUR
391-0480
Spreaders, L-shaped, Sterilin®
Uitstrijkspatels Celspatels
Designed for spreading and dispersing liquids across the surface of agar culture plates. L-shaped with smooth rounded surfaces to prevent gouging of agar during inoculation.