Transfer pipettes, Sterilin®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sterilin®
SLIN200CEA 59.1 EUR
SLIN200C SLIN202C 734-0460
Transfer pipettes, Sterilin®
Pipetten Pasteur-/transferpipetten
These disposable pipettes, manufactured from non toxic LDPE, transfer liquids safely and accurately. Ideal for general laboratory use, sample storage and transport.
  • Integral filling bulb eliminates contamination associated with separate rubber bulbs
  • Excellent transparency and uniform thickness
  • Can be heat sealed for sample storage and transportation
  • Heat-sealed bulbs suitable for storage in the gas phase of liquid nitrogen
  • 1 ml graduated version: Suitable for general laboratory use, storage and transportation of samples
  • 3 ml graduated version: Suitable for blood banking and general laboratory use
  • Narrow stem version: Suitable for use as a freeze vial, sedimentation and transport