Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
U heeft gezocht op: "draagtassen"
https://nl.vwr.com/stibo/smallweb/std.lang.all/49/95/18944995.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"10"
  List view Grid view Easy View (new)
Sorteer op:
Catalogus nummer: (BRDY050979)
Blocked
Leverancier:  BRADY & CO LTD, W. H.
Omschrijving:   Tas gemaakt van nylon met bevestigingslus aan achterkant.
UOM:  1 * 1 ST
Catalogus nummer: (BRDY051172)
Leverancier:  BRADY & CO LTD, W. H.
Omschrijving:   Pouches for lockout devices with loops for hanging on belt.
UOM:  1 * 1 ST
Catalogus nummer: (BRDY099162)
Leverancier:  BRADY & CO LTD, W. H.
Omschrijving:   These duffle bags are made of 600 denier polyester that are used for lock out kits.
UOM:  1 * 1 ST
Catalogus nummer: (BRDY806200)
Leverancier:  BRADY & CO LTD, W. H.
Omschrijving:   Lockout satchel is made from 1000 denier red cordura nylon with a 95 cm shoulder strap.
UOM:  1 * 1 ST
Catalogus nummer: (PRTWB901BKR)
Leverancier:  Portwest Clothing
Omschrijving:   The bag made of highly durable 600D polyester fabric with a side zip pocket for stacking boots or helmet.
UOM:  1 * 1 ST
Catalogus nummer: (113-6941)
Leverancier:  Portwest Clothing
Omschrijving:   Medium size travel bag
UOM:  1 * 1 ST
Leverancier:  Portwest Clothing
Omschrijving:   This polyester backpack is ideal for workers, cyclists or school children who use highly reflective accessories to make them more visible to motorists...
UOM:  1 * 1 ST
Catalogus nummer: (KWIN100016240)
Leverancier:  FRISTADS KANSAS GROUP
Omschrijving:   Tool bag made of heavy-duty split leather.
UOM:  1 * 1 ST
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 10
  1  2  Volgende