Netherlands
Order Entry
Accessories for Beta Easy Lock Autoclaves, PBI
Supplier:  VWR Collection
Certificates
undefined
Accessories for Beta Easy Lock Autoclaves, PBI
Product Information
  • Catalog #
    For
    Pk
    Availability
    Price
  • Data printer
    Data printer
    481-0673
    Autoclave Beta Easy-Lock
    1
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 395,00 €
    395,00 €
    1 * 1 ST
       395,00 €
    Specifications
    Environmentally Preferable
  • Ecobox-25 basket: (Ø×H): 240×420 mm, in stainless steel AISI 304 (max. load: 1 basket)
    Ecobox-25 basket: (Ø×H): 240×420 mm, in stainless steel AISI 304 (max. load: 1 basket)
    710-0887
    Autoclaves, ECO 25
    1
    1 * 1 ST
    1 * 1 ST - 238,00 €
    238,00 €
    1 * 1 ST
       238,00 €
    Specifications
    Environmentally Preferable
  • Ecobox-25-mini basket: (Ø×H): 240×210 mm, in stainless steel AISI 304 (max. load: 2 baskets)
    Ecobox-25-mini basket: (Ø×H): 240×210 mm, in stainless steel AISI 304 (max. load: 2 baskets)
    481-0669
    Autoclaves, ECO 25
    1
    1 * 1 ST
    1 * 1 ST - 146,00 €
    146,00 €
    1 * 1 ST
       146,00 €
    Specifications
    Environmentally Preferable
  • Ecobox-50 basket: (Ø×H): 310×285 mm, in stainless steel AISI 304 (max. load: 2 baskets)
    Ecobox-50 basket: (Ø×H): 310×285 mm, in stainless steel AISI 304 (max. load: 2 baskets)
    481-0672
    Autoclaves, ECO 50
    1
    1 * 1 ST
    1 * 1 ST - 231,00 €
    231,00 €
    1 * 1 ST
       231,00 €
    Specifications
    Environmentally Preferable
  • Ecobox-50-mini basket: (Ø×H): 310×190 mm in stainless steel AISI 304 (max. load: 3 baskets)
    Ecobox-50-mini basket: (Ø×H): 310×190 mm in stainless steel AISI 304 (max. load: 3 baskets)
    481-0668
    Autoclaves, ECO 50
    1
    1 * 1 ST
    1 * 1 ST - 160,00 €
    160,00 €
    1 * 1 ST
       160,00 €
    Specifications
    Environmentally Preferable
  • IQ-OQ-PQ instruction manual for Beta 25 Easy Lock autoclave
    IQ-OQ-PQ instruction manual for Beta 25 Easy Lock autoclave
    481-0724
    Autoclaves, ECO 25
    1
    1 * 1 ST
    1 * 1 ST - 548,00 €
    548,00 €
    1 * 1 ST
       548,00 €
    Specifications
    Environmentally Preferable
  • Betabox basket: Ext. Ø 310×380 mm, in stainless steel AISI 304 (max. load: 1 basket)
    Betabox basket: Ext. Ø 310×380 mm, in stainless steel AISI 304 (max. load: 1 basket)
    481-0677
    Autoclaves, ECO 50
    1
    1 * 1 ST
    1 * 1 ST - 44,30 €
    44,30 €
    1 * 1 ST
       44,30 €
    Specifications
    Environmentally Preferable