Column head with valve stopcock acc. to Antlinger
Leverancier: Witeg
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WITG1.610.029EA 687.08 EUR
WITG1.610.029
Column head with valve stopcock acc. to Antlinger
Onderdelen voor destillatie Destillatiekoppen
Borosilicate glass3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Columns heads acc. to Antlinger, with PTFE-fine-dosing valve, cone to cloumn NS 29/32.