Restraint rope lanyards
Leverancier: Miller by Honeywell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DALL1002883EA 29.63 EUR
DALL1002883 DALL1004711 DALL1002882 DALL1002884
Restraint rope lanyards
Lightweight, versatile restraint and fall arrest lanyards, made from polyamide, designed for maximum protection.
  • Adapted rope for work requiring limited movement and where there is a sufficient potential fall distance
  • Ropes come with a buckle and a karabiner