1-Boc-4-piperidone
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
XXXXPZ1563EA
0
EUR
XXXXPZ1563
1-Boc-4-piperidone
1-Boc-4-piperidone
Formule:
C₁₀H₁₇NO₃ MW: 199,25 g/mol |
MDL Nummer:
MFCD00151800 CAS nummer: 79099-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...