2'-Fluorobiphenyl-4-carboxylic acid ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H33301.06EA
192
EUR
H33301.06
2'-Fluorobiphenyl-4-carboxylic acid ≥97%
2'-Fluorobiphenyl-4-carboxylic acid
Formule:
C₁₃H₉FO₂ MW: 216,21 g/mol Smeltpunt: 230…232 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00278732 CAS nummer: 365-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...