1-(2-Methylbenzyl)piperazine ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H55203.03EA
27.66
EUR
H55203.03
1-(2-Methylbenzyl)piperazine ≥97%
1-(2-Methylbenzyl)piperazine
Formule:
C₁₂H₁₈N₂ MW: 190,29 g/mol Kookpunt: 77…80 °C (0,1 mmHg) Dichtheid: 0,988 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD02256026 CAS nummer: 5321-47-1 EINECS: 226-183-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...