Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
1-(4-Bromophenyl)cyclopropanecarbonitrile
1-(4-Bromophenyl)cyclopropanecarbonitrile
1-(4-Bromophenyl)cyclopropanecarbonitrile
1-(4-Bromophenyl)cyclopropanecarbonitrile
Supplier:
Thermo Scientific
Certificates
Expand image
1-(4-Bromophenyl)cyclopropanecarbonitrile
Supplier:
Thermo Scientific
About this item
CAS Number:
124276-67-1
:
Formula:
C₁₀H₈BrN
MDL Number:
MFCD01314308
:
222.0812 g/mol
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
MAYBRJC03711.1
RJC03711.1
Glass bottle for solids
1 g
Log in
for availability
Log in
for availability
1 * 1 g
-
+
Inquire For Price
1 * 1 g
-
+
Inquire For Price
MAYBRJC03711.10
RJC03711.10
Glass bottle for solids
10 g
Log in
for availability
Log in
for availability
1 * 10 g
-
+
Inquire For Price
1 * 10 g
-
+
Inquire For Price