Calcium chloride cylinders
Leverancier: Witeg
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WITG5.624.104EA 168.66 EUR
WITG5.624.104
Calcium chloride cylinders
Exsiccatoren Exsiccatoren, glas
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes