Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Leak diverter for roofs
Leverancier: New Pig
PIG®
A range of different sizes and shapes of leak diverter to protect workers equipment and floors from overhead leaks.
  • Diverter uses a 19 mm drainage hose connection to drain water safely away from your work area
  • Non permeable tarpaulin is vinyl coated for strength and durability; resists punctures, UV and mildew for long life
  • Heavy-duty grommets for easy installation and tear prevention
  • Flow rate is 12,6 litres/minute when used with a 12 mm ID clear drainage hose
Bestelinformatie: Please order hoses and hanging straps separately.
Order
Expand All / Collapse All
Omschrijving l×b VPE VWR catalogus nummer Hoeveelheid
Lekkageomleider, voor daken, doorzichtig, vuur-, knel- en scheurbestendig 760×760 mm 1 121-0168
Leak diverter, clear, 8 grommets 1520×1520 mm 1 NEWPTLS462-CL
Lekkageomleider, voor daken, doorzichtig, vuur-, knel- en scheurbestendig 1520×1520 mm 1 121-0159
Lekkageomleider, voor daken, doorzichtig, vuur-, knel- en scheurbestendig 3050×1520 mm 1 121-0169
Lekkageomleider, voor daken, doorzichtig, vuur-, knel- en scheurbestendig 6100×3050 mm 1 121-0170
Lekkageomleider, voor daken, doorzichtig, vuur-, knel- en scheurbestendig 3660× 3660 mm 1 121-0162
Lekkageomleider, voor daken, doorzichtig, vuur-, knel- en scheurbestendig 4570×4570 mm 1 121-0163
Lekkageomleider, voor daken, doorzichtig, vuur-, knel- en scheurbestendig 6100×6100 mm 1 121-0164
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Accessoires
Expand All / Collapse All
Accessories
Omschrijving VPE VWR catalogus nummer Hoeveelheid
Hangende rubberen banden, lengte 355,5 mm 4 121-0165
Hangende rubberen banden, lengte 482,5 mm 4 121-0166
Hangende rubberen banden, lengte 609,5 mm 4 121-0167
Heldere afvoerslang, lengte 3 m 1 121-0172
Heldere afvoerslang, lengte 7,32 m 762 cm 121-0171
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid
Een nieuwe uitstraling en een hernieuwde belofte
Gebouwd op de kenmerken & middelen die u kent, geoptimaliseerd met de innovatie van Avantor ®. Ons nieuwe merk symboliseert flexibiliteit en ons vermogen om ons aan de behoeften van de klant aan te passen.
KOM MEER TE WETEN BEGIN MET WINKELEN