Netherlands
Order Entry
Fluid aspiration system, BioChem VacuuCenter, BVC basic series
Fluid aspiration system, BioChem VacuuCenter, BVC basic series
Supplier:  VACUUBRAND
   Sustainable Options Available
Certificates
undefined
Fluid aspiration system, BioChem VacuuCenter, BVC basic series
Supplier:  VACUUBRAND
   Sustainable Options Available
Specifications
  • Catalog No:
  • 181-0299
  • 181-0298
  • Model:
  • BioChem-VacuuCenter BVC basic G
  • BioChem-VacuuCenter BVC basic
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Vacuum pump:
  • supplied without pump
  • supplied without pump
  • Final pressure of vacuum pump:
  • 150 mbar
  • 150 mbar
  • Protection class:
  • IP 54
  • IP 54
  • Weight:
  • 2,8 kg
  • 2,8 kg
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    W×D×H
    Pk
    Availability
    Price
  • BioChem-VacuuCenter BVC basic: with 4 l PP collection bottle for general aspiration tasks, completely autoclavable together with a 0,2 µm protection filter
    BioChem-VacuuCenter BVC basic: with 4 l PP collection bottle for general aspiration tasks, completely autoclavable together with a 0,2 µm protection filter
    181-0298
    20727000
    250×200×490 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
    Supplier Article Number: 20727000
    Local Article Number: VACU20727000
    Specifications:
      Energy Efficient
    More
    Accessories
    Specifications
    Model
    BioChem-VacuuCenter BVC basic
    Environmentally Preferable
    Energy Efficient
    Vacuum pump
    supplied without pump
    Final pressure of vacuum pump
    150 mbar
    Protection class
    IP 54
    Weight
    2,8 kg
      Energy Efficient
  • BioChem-VacuuCenter BVC basic G: with 2 l borosilicate glass collection flask with splinter-proof coating and leakage protection, with high chemical resistance for aggressive disinfectants like chlorine bleach; completely autoclavable together with 0,2 µm protection filter.
    BioChem-VacuuCenter BVC basic G: with 2 l borosilicate glass collection flask with splinter-proof coating and leakage protection, with high chemical resistance for aggressive disinfectants like chlorine bleach; completely autoclavable together with 0,2 µm protection filter.
    181-0299
    20727100
    250×200×490 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 150,00 €
    1 150,00 €
    1 * 1 ST
       1 150,00 €
    Supplier Article Number: 20727100
    Local Article Number: VACU20727100
    Specifications:
      Energy Efficient
    More
    Accessories
    Specifications
    Model
    BioChem-VacuuCenter BVC basic G
    Environmentally Preferable
    Energy Efficient
    Vacuum pump
    supplied without pump
    Final pressure of vacuum pump
    150 mbar
    Protection class
    IP 54
    Weight
    2,8 kg
      Energy Efficient