Combined conductivity electrode, MultiSens
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
662-1758EA 905 EUR
662-1758
Combined conductivity electrode, MultiSens
Elektroden Geleidbaarheidselektroden
Multifunctional 4-pole conductivity cell with integrated temperature sensor (Pt1000).
  • Screw plug head with thread Pg 13,5
  • Four platinum rings
Pressure range: 6 bar
Leveringsinformatie: Supplied with washer 9980/52 and O-ring type 9932/07. Connecting cable needs to be ordered separately.