Netherlands
Order Entry
Desiccator cabinets, anti-static, SICCO maxi series
Desiccator cabinets, anti-static, SICCO maxi series
Supplier:  Bohlender
Certificates
undefined
Desiccator cabinets, anti-static, SICCO maxi series
Supplier:  Bohlender
Product Information
  • Catalog #
    Supplier #
    Shelves
    With
    W×D×H ext.
    W×D×H int.
    Pk
    Availability
    Price
  • Maxi 2 Antistatic (140 L)
    Maxi 2 Antistatic (140 L)
    BOHLV1923-07
    V1923-07
    2
    2 chambers and 2 doors, 17 shelf positions, 1× tray and hygrometer supplied per chamber
    560×580×1150 mm
    495×540×500 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 140,00 €
    8 140,00 €
    1 * 1 ST
       8 140,00 €
    Supplier Article Number: V1923-07
    Local Article Number: BOHLV1923-07
    Specifications:
    Specifications
    Environmentally Preferable
  • Maxi 1 Antistatic (280 L)
    Maxi 1 Antistatic (280 L)
    BOHLV1922-07
    V1922-07
    4
    1 chamber and 1 door, 34 shelf positions, 1× tray and hygrometer supplied
    560×580×1150 mm
    495×540×1030 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 200,00 €
    6 200,00 €
    1 * 1 ST
       6 200,00 €
    Supplier Article Number: V1922-07
    Local Article Number: BOHLV1922-07
    Specifications:
    Specifications
    Environmentally Preferable
Frequently Bought Together