Cold trap with scealing cap
Leverancier: KNF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KNFA057955EA 0 EUR
KNFA057955
Cold trap with scealing cap
Cold trap made of borosilicate glass 3.3, with GL 45 closing cap.
  • Features hose with threaded caps GL 18