Upright microscopes, BA210 Elite and BA310 Elite
Supplier: MOTIC
Certificates
About this item
BA210 Elite gives high performance in educational environments for life sciences and medical applications.
BA310 Elite is dedicated to demanding applications in universities, clinics and other life science or medical applications requiring quality optical performance. Full Köhler configuration provides maximum illumination quality.
Delivery information: Supplied with plan achromatic objectives (4), blue filter, immersion oil (5 ml), power cord with hanger, dust cover, an Allen key and spare fuse.
Specifications
- Catalog No:
- 630-1826
- 630-1825
- MOTI1100100402966
- MOTI1100100403221
- Model:
- BA310 Elite
- BA310 Elite
- BA310 Elite
- BA310 Elite
- Observation method:
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Head:
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- Eyepieces:
- WF 10×/20 mm with pointer adjustment
- WF 10×/20 mm with pointer adjustment
- WF 10×/20 mm with pointer adjustment
- WF 10×/20 mm with pointer adjustment
- Nosepiece:
- Quintuple, reversed
- Quintuple, reversed
- Quintuple, reversed
- Quintuple, reversed
- Stage:
- 180×170 mm (movement: 80×55 mm)
- 180×170 mm (movement: 80×55 mm)
- 180×170 mm (movement: 80×55 mm)
- 180×170 mm (movement: 80×55 mm)
- Condenser:
- Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
- Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
- Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
- Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
- Focusing system:
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Illumination:
- Halogen interchangeable with LED module
- Halogen interchangeable with LED module
- Halogen interchangeable with LED module
- Halogen interchangeable with LED module
Specifications
Product Information
- Catalog #Supplier #ModelTypeMagnificationPkAvailabilityPrice
- 630-18251100100402433BA310 EliteBinocular4×, 10×, 40×, 100×1Log in for availability1 * 1 ST1 469,00 €Supplier Article Number: 1100100402433Local Article Number: MOTI1100100402433Specifications:
More
Accessories
Specifications
ModelBA310 EliteEnvironmentally PreferableObservation methodBrightfieldHead30° inclined, 360° rotatingEyepiecesWF 10×/20 mm with pointer adjustmentNosepieceQuintuple, reversedStage180×170 mm (movement: 80×55 mm)CondenserAbbe N.A. 0,90/1,25 with iris diaphragm and slider slotFocusing systemCoaxial coarse and fine knobsIlluminationHalogen interchangeable with LED module - MOTI11001004032211100100403221BA310 EliteBinocular4×, 10×, 40×1Log in for availability1 * 1 ST1 328,00 €Supplier Article Number: 1100100403221Local Article Number: MOTI1100100403221Specifications:
More
Accessories
Specifications
ModelBA310 EliteEnvironmentally PreferableObservation methodBrightfieldHead30° inclined, 360° rotatingEyepiecesWF 10×/20 mm with pointer adjustmentNosepieceQuintuple, reversedStage180×170 mm (movement: 80×55 mm)CondenserAbbe N.A. 0,90/1,25 with iris diaphragm and slider slotFocusing systemCoaxial coarse and fine knobsIlluminationHalogen interchangeable with LED module - 630-18261100100402443BA310 EliteTrinocular4×, 10×, 40×, 100×1Log in for availability1 * 1 ST1 686,00 €Supplier Article Number: 1100100402443Local Article Number: MOTI1100100402443Specifications:
More
Accessories
Specifications
ModelBA310 EliteEnvironmentally PreferableObservation methodBrightfieldHead30° inclined, 360° rotatingEyepiecesWF 10×/20 mm with pointer adjustmentNosepieceQuintuple, reversedStage180×170 mm (movement: 80×55 mm)CondenserAbbe N.A. 0,90/1,25 with iris diaphragm and slider slotFocusing systemCoaxial coarse and fine knobsIlluminationHalogen interchangeable with LED module - MOTI11001004029661100100402966BA310 EliteTrinocular4×, 10×, 40×1Log in for availability1 * 1 ST1 545,00 €Supplier Article Number: 1100100402966Local Article Number: MOTI1100100402966Specifications:
More
Accessories
Specifications
ModelBA310 EliteEnvironmentally PreferableObservation methodBrightfieldHead30° inclined, 360° rotatingEyepiecesWF 10×/20 mm with pointer adjustmentNosepieceQuintuple, reversedStage180×170 mm (movement: 80×55 mm)CondenserAbbe N.A. 0,90/1,25 with iris diaphragm and slider slotFocusing systemCoaxial coarse and fine knobsIlluminationHalogen interchangeable with LED module
Objectives
Product Information
- Catalog #SeqDescriptionMagnificationPkAvailabilityPrice
- Supplier Article Number: 1101001702911Local Article Number: MOTI1101001702911
Camera Adapters
Product Information
- Catalog #SeqDescriptionPkAvailabilityPrice
- Supplier Article Number: 1101001901781Local Article Number: MOTI1101001901781
Camera
Product Information
- Catalog #PkAvailabilityPrice
- Supplier Article Number: 1101001906191Local Article Number: MOTI1101001906191