Freezers, built in
Leverancier: Liebherr
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LIEBIGS1113EA 0 EUR
LIEBIGS1113 LIEBEG1113 LIEBIG1156 LIEBIG956 LIEBIGN2566 LIEBIG1956
Freezers, built in
Vriezers Vriezers voor laboratoria en algemeen gebruik
Freezers are designed with the precision control system and MagicEye which ensures accurate temperature control.
  • Simple and convenient to operate
  • Automatic SuperFrost function freezes food rapidly while preserving the vitamins
  • Plumbed-in IceMaker makes top-quality ice cubes for all occasions with switched off options
The transparent freezer drawers are removable giving flexible storage solutions as well as providing a perfect view of the stored food. The drawers in the FrostSafe system are sealed all the way round, which maintains the low temperature inside the appliance and enhances its energy-efficient operation. All drawers and glass shelves can be easily removed from the freezer compartments in all fridge freezers by VarioSpace concept, creates extra storage space.
NoFrost option provide professional-standard cooling performance. Chilled circulating air freezes the food and humidity is expelled so that the freezer is always ice-free and produce never frosts over. SmartFrost option vastly reduces ice build-up in the interior and on frozen foods so that defrosting is required far less often. The interior walls are smooth and easy to clean.