Netherlands
Order Entry
Joint clips for conical joints, POM, Quickfit®
Joint clips for conical joints, POM, Quickfit®
Supplier:  SciLabware
Certificates
undefined
Joint clips for conical joints, POM, Quickfit®
Supplier:  SciLabware
Product Information
  • Catalog #
    Supplier #
    Colour
    For NS
    Pk
    Availability
    Price
  • 201-8185
    KC10
    Mint green
    10/19
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 48,20 €
    48,20 €
    1 * 10 ST
       48,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-8186
    KC12
    Purple
    12/21
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 49,20 €
    49,20 €
    1 * 10 ST
       49,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-8566
    KC14
    Yellow
    14/23
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52,20 €
    52,20 €
    1 * 10 ST
       52,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-8567
    KC19
    Blue
    19/26
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52,20 €
    52,20 €
    1 * 10 ST
       52,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-8568
    KC24
    Green
    24/29
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 56,20 €
    56,20 €
    1 * 10 ST
       56,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-8569
    KC29
    Red
    29/32
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 58,20 €
    58,20 €
    1 * 10 ST
       58,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-8570
    KC34
    Orange
    34/35
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 96,70 €
    96,70 €
    1 * 10 ST
       96,70 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-3032
    KC40
    Ochre
    40/38
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,20 €
    72,20 €
    1 * 10 ST
       72,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 201-8192
    KC45
    Amber
    45/40
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 76,20 €
    76,20 €
    1 * 10 ST
       76,20 €
    Specifications:
    Specifications
    Environmentally Preferable