Cleanroom goggles, Coverall Autoclave
Leverancier: Bollé Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Coverall Autoclave
BOLECOVACLAVEEA 19.51 EUR
BOLECOVACLAVE
Cleanroom goggles, Coverall Autoclave
Ruimzichtbrillen
These cleanroom Coverall goggles with interchangeable ventilation ensure protection and comfort. It is possible to wear prescription spectacles under them.
  • PLATINUM coated lens: Coating on both sides ensures high scratch-resistance (1,4 cd/m²), high resistance to the most aggressive chemicals and slows the appearance of fogging
  • Adjustable strap
  • Upper indirect and interchangeable ventilation
  • Ultra-comfort autoclavable goggles
Weight: 54 g
Certificaten: EN 166