Distillation flasks, with side arm
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIBB1280/14MEA 1080 EUR
BIBB1280/14M
Distillation flasks, with side arm
Pyrex® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For general distillation work. Side-arm is inclined at 75° to the vertical axis. For use with thermometers calibrated for 76 mm immersion. Side-arm length is 140 mm.