Reusable injection syringes, ULTRA-ASEPT®
Leverancier: Henke-Sass, Wolf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ULTRA-ASEPT®
HSWA2450030100EA 0 EUR
HSWA2450030100
Reusable injection syringes, ULTRA-ASEPT®
Spuiten Injectiespuiten, medisch
Non sterile, reusable, glass barrel and stainless steel piston. These syringes can be used for human and veterinary medicine.
  • Interchangeable barrel
  • Completely field serviceable
  • Ground-in piston