Pre-weighted calibration kits, EasiVial™
Supplier: VARIAN
Certificates
About this item
The quickest and most convenient method to deliver an accurate 12 point column calibration for organic and aqueous GPC/SEC column calibration.
- Pre-prepared, time saving product for rapid and reliable GPC column calibration
- Eliminates tedious weighing procedures to improve calibration accuracy
- Reduces solvent dispensing to limit risks associated with handling solvents
- For conventional and multi detector GPC to maximize applicability
The mass of each polymer in the vial is accurately known, so that upon addition of a fixed volume of eluent, the solution is prepared at a precise concentration. It is ideal for both conventional and multi detector GPC calibration. Simple to prepare and manually inject or transfer to autosampler vials or place directly into a compatible autosampler.
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- EasiVial PS-M, 2 mlSupplier Article Number: PL2010-0301Local Article Number: VARIPL2010-0301
- EasiVial PS-M, 4 mlSupplier Article Number: PL2010-0300Local Article Number: VARIPL2010-0300
- EasiVial PS-L, 2 mlSupplier Article Number: PL2010-0401Local Article Number: VARIPL2010-0401
- EasiVial PS-L, 4 mlSupplier Article Number: PL2010-0400Local Article Number: VARIPL2010-0400
- EasiVial PM, 2 mlSupplier Article Number: PL2020-0201Local Article Number: VARIPL2020-0201
- EasiVial PM, 4 mlSupplier Article Number: PL2020-0200Local Article Number: VARIPL2020-0200
- EasiVial PEG, 2 mlSupplier Article Number: PL2070-0201Local Article Number: VARIPL2070-0201
- EasiVial PEG, 4 mlSupplier Article Number: PL2070-0200Local Article Number: VARIPL2070-0200
- EasiVial PS-H, 4 mlSupplier Article Number: PL2010-0200Local Article Number: VARIPL2010-0200
- EasiVial PEG/PEO, 4 mlSupplier Article Number: PL2080-0200Local Article Number: VARIPL2080-0200
- PS-H Tri-Pack, 2 mlSupplier Article Number: PL2010-0202Local Article Number: VARIPL2010-0202
- PS-H Tri-Pack, 4 mlSupplier Article Number: PL2010-0203Local Article Number: VARIPL2010-0203
- PS-L Tri-Pack, 3 mlSupplier Article Number: PL2010-0402Local Article Number: VARIPL2010-0402
- PS-L Tri-Pack, 4 mlSupplier Article Number: PL2010-0403Local Article Number: VARIPL2010-0403
- PMMA Tri-Pack, 2 mlSupplier Article Number: PL2020-0202Local Article Number: VARIPL2020-0202
- PEG Tri-Pack, 2 mlSupplier Article Number: PL2070-0202Local Article Number: VARIPL2070-0202
- PEG/PEO Tri-Pack, 2 mlSupplier Article Number: PL2080-0202Local Article Number: VARIPL2080-0202
- PEG/PEO Tri-Pack, 4 mlSupplier Article Number: PL2080-0203Local Article Number: VARIPL2080-0203