Netherlands
Order Entry
Human Synthetic GALR1 (from HEK293 Cells)
Human Synthetic GALR1 (from HEK293 Cells)
New Product
Human Synthetic GALR1 (from HEK293 Cells)
New Product
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P47211
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    GAL1-R|Galanin receptor type 1|GALNR|GALNR1|GALR-1
  • Protein/peptide name:
    GALR1
  • Molecular weight:
    Full length human GALR1 protein has a MW of 39 kDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilization buffer (20 mM tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  •  
    Pack type
  • ANTIA324752-10
    A324752-10
    Vial
    1 * 1 EA
    1 * 1 EA - 1 280,00 €
    1 280,00 €
    1 * 1 EA
       1 280,00 €
     : A324752-10
     : ANTIA324752-10
    New Product
    New Product
  • ANTIA324752-50
    A324752-50
    Vial
    1 * 1 EA
    1 * 1 EA - 5 910,00 €
    5 910,00 €
    1 * 1 EA
       5 910,00 €
     : A324752-50
     : ANTIA324752-50
    New Product
    New Product
  • ANTIA324752-100
    A324752-100
    Vial
    1 * 1 EA
    1 * 1 EA - 10 680,00 €
    10 680,00 €
    1 * 1 EA
       10 680,00 €
     : A324752-100
     : ANTIA324752-100