Netherlands
Order Entry
HIV-1 Recombinant HIV-1 Gag p15 (from E. coli)
HIV-1 Recombinant HIV-1 Gag p15 (from E. coli)
Supplier:  Antibodies.com
New Product
Certificates
HIV-1 Recombinant HIV-1 Gag p15 (from E. coli)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    HIV-1
  • Storage conditions:
    Upon delivery aliquot and store at −20 °C . Avoid freeze/thaw cycles.
  • Carrier-free:
    N
  • Protein/peptide name:
    HIV-1 Gag p15
  • Purity:
    >90% (by SDS-PAGE)
  • Molecular weight:
    15 kDa
  • Concentration:
    0.42 mg/ml
  • Formulation:
    Supplied in 20 mM Tris-HCl, pH 7,5, with 50 mM NaCl, 10 mM Mercaptoethanol and 50% Glycerol.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE, WB, DB, ELISA

Specifications

Product Information

  • Catalog #
    Supplier #
    Pack type
    Availability
    Price
  • ANTIA468-20
    A468-20
    Vial
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 405,00 €
    405,00 €
    1 * 1 EA
       405,00 €

    Supplier Article Number: A468-20
    Local Article Number: ANTIA468-20
    Specifications:
    New Product
    New Product
  • ANTIA468-100
    A468-100
    Vial
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 1 210,00 €
    1 210,00 €
    1 * 1 EA
       1 210,00 €

    Supplier Article Number: A468-100
    Local Article Number: ANTIA468-100
    Specifications:
    New Product
    New Product