Netherlands
Order Entry
Anti-IL-19 Rabbit Monoclonal Antibody [clone: EPNCIR168]
Anti-IL-19 Rabbit Monoclonal Antibody [clone: EPNCIR168]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-IL-19 Rabbit Monoclonal Antibody [clone: EPNCIR168]
Supplier:  Abcam
New Product

Product Information

  • Catalog #
    Supplier #
    Size
    Pack type
    Availability
    Price
  • ABCAAB154187-100
    AB154187-100
    100 µl
    Vial
    Log in for availability
    Log in for availability
    1 * 100 µl
    1 * 100 µl - 663,00 €
    663,00 €
    1 * 100 µl
       663,00 €

    Supplier Article Number: AB154187-100
    Local Article Number: ABCAAB154187-100
    Specifications:
    Specifications
    Environmentally Preferable
  • ABCAAB154187-1ML
    AB154187-1ML
    1 ml
    Vial
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 5 970,00 €
    5 970,00 €
    1 * 1 mL
       5 970,00 €

    Supplier Article Number: AB154187-1ML
    Local Article Number: ABCAAB154187-1ML
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product