Netherlands
Order Entry
LN₂ Supply Tanks
LN₂ Supply Tanks
New Product
Certificates
undefined
LN₂ Supply Tanks
New Product
Product Information
  • Catalog #
    Supplier #
    Availability
    Price
  • LN₂ supply tank, 50 L, 902H×406D mm
    LN₂ supply tank, 50 L, 902H×406D mm
    BARN8127
    8127
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9 090,00 €
    9 090,00 €
    1 * 1 ST
       9 090,00 €
    Supplier Article Number: 8127
    Local Article Number: BARN8127
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • LN₂ supply tank, 180 L, 1613H×508D mm
    LN₂ supply tank, 180 L, 1613H×508D mm
    BARN8120
    8120
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13 270,00 €
    13 270,00 €
    1 * 1 ST
       13 270,00 €
    Supplier Article Number: 8120
    Local Article Number: BARN8120
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product